Learn More
Cayman Chemical sn-Glycerol-3-phosphate (cyclohexyl ammonium salt hydrate), Purity: ≥95%, 100 mg
SDP
Catalog No. 502984925
Encompass_Preferred
A lipid biosynthesis intermediate; functions in oxidative phosphorylation as part of the glycerophosphate shuttle; used in synthetic biology, studies of reactive oxygen species, and to measure phosphatidic acid in cell cultures; vital for systemic acquired resistance in plants; levels in saliva are altered in patients with Alzheimer’s, dementia, and Lewy body disease
Catalog No. 50-298-4925 Supplier Cayman Chemical Supplier No. 20729100 MG
A lipid biosynthesis intermediate; functions in oxidative phosphorylation as part of the glycerophosphate shuttle; used in synthetic biology, studies of reactive oxygen species, and to measure phosphatidic acid in cell cultures; vital for systemic acquired resistance in plants; levels in saliva are altered in patients with Alzheimer’s, dementia, and Lewy body disease