Learn More
AccuStandard, Inc SNION STD-NITRATE AS NITRO 500
SDP
Catalog No. NC9316236
Encompass
Anion Standard - Nitrate as Nitrogen. 100 mL. 100 ?g/mL in Water.
Catalog No. NC9316236 Supplier AccuStandard, Inc Supplier No. ICNO3N10X5
Anion Standard - Nitrate as Nitrogen. 100 mL. 100 ?g/mL in Water.