Learn More
Grainger Snip Tip Applicator 3.7 oz Plastic PK100
Catalog No. 502764076
Encompass_Preferred
The ECLECTIC Snip Tip Applicator Tube is a plastic tube with a tapered tip and cap for precise glue application. It has a 0.34 fl oz capacity and is 5" long with a 0.1875" opening. The tubes are sold in packs of 12 and made in the USA. They can dispense adhesives, epoxies, silicones, lubricants, and more. The tapered tip allows for precise application in tight spaces. The cap helps keep the contents fresh between uses. Overall, this is a 12-pack of small plastic applicator tubes for accurately dispensing various liquids.
Catalog No. 50-276-4076 Supplier Grainger Supplier No. 16NH02
The ECLECTIC Snip Tip Applicator Tube is a plastic tube with a tapered tip and cap for precise glue application. It has a 0.34 fl oz capacity and is 5" long with a 0.1875" opening. The tubes are sold in packs of 12 and made in the USA. They can dispense adhesives, epoxies, silicones, lubricants, and more. The tapered tip allows for precise application in tight spaces. The cap helps keep the contents fresh between uses. Overall, this is a 12-pack of small plastic applicator tubes for accurately dispensing various liquids.