Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SNIPER(BRD)-1 | 2095244-54-3 | 99.7% | 1056.73 | 1 MG
SDP

Catalog No. 5000438384
Encompass_Preferred

SNIPER(BRD)-1 is a SNIPER degrader of BRD4, cIAP1, and XIAP. It has IC50 values of 6.8 nM, 17 nM, and 49 nM for cIAP1, cIAP2, and XIAP, respectively. It can be used for cancer research.

  • SNIPER degrader of BRD4, cIAP1, and XIAP.
  • IC50 values: cIAP1: 6.8 nM, cIAP2: 17 nM, XIAP: 49 nM.
  • Can be used for cancer research.
  • Significantly reduces protein levels of BRD4, cIAP1, and XIAP in LNCaP cells.
  • Purity: 99.68%.
  • Appearance: solid, off-white to light yellow.

Catalog No. 50-004-38384 Supplier Medchemexpress LLC Supplier No. HY1118751MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SNIPER(BRD)-1 is a SNIPER degrader of BRD4, cIAP1, and XIAP. It has IC50 values of 6.8 nM, 17 nM, and 49 nM for cIAP1, cIAP2, and XIAP, respectively. It can be used for cancer research.

  • SNIPER degrader of BRD4, cIAP1, and XIAP.
  • IC50 values: cIAP1: 6.8 nM, cIAP2: 17 nM, XIAP: 49 nM.
  • Can be used for cancer research.
  • Significantly reduces protein levels of BRD4, cIAP1, and XIAP in LNCaP cells.
  • Purity: 99.68%.
  • Appearance: solid, off-white to light yellow.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.