Learn More
Sentry Air Systems 6" dia. Flex Hose Assembly
SDP
Catalog No. NC2589906
Encompass
6" diameter and 72" long flexible hose with round hood and clamp for Model 400 units. Made of flame retardant material.
Catalog No. NC2589906 Supplier Sentry Air Systems Supplier No. SS006BH
6" diameter and 72" long flexible hose with round hood and clamp for Model 400 units. Made of flame retardant material.