Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Snow Polymer Demonstration Kit
SCI_ED

Catalog No. S06974 Shop All Aldon Products
Change view
Click to view available options
:
Snow Polymer Demonstration Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S06974 Snow Polymer Demonstration Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06974 Supplier Innovating Science™ Supplier No. IS7043

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

Includes

  • Snow polymer (25 g)
  • Cups (5)
  • Stir sticks (5)

Ideal for classroom demonstration to learn super absorbent properties of a polymer.

  • Hydrophilic polymer instantly absorbs water
  • Expands to over 40 times its original volume
  • End result is fluffy flakes of fake snow

Order Info

Performs five demos.

Specifications

Product Type Snow Polymer Demonstration Kit
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) For classroom demonstration to learn super absorbent properties of a polymer
Includes 25 g snow polymer, 5 cups, 5 stir sticks
Quantity 1
Dimensions (L x W x H) 4.25 x 4.625 x 5.5 in.
Class Size 5 Demonstrations
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.