Learn More
Eurofins Discoverx SNRK, active, 10 µg. (14-932)
Supplier:  Eurofins Discoverx 14932
Encompass
C-terminal 6xHis-tagged recombinant, human SNRK amino acids 1-505. SNF-related serine/threonine-protein kinase may play a role in hematopoietic cell proliferation or differentiation. It is a potential mediator of neuronal apoptosis. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9437