Learn More
Aviva Systems Biology Corp SNRNP200 Antibody - N-terminal region (ARP36414_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36414_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SNRNP200. It was validated on Western Blot. . Alternate names: ASCC3L1, BRR2, FLJ11521, HELIC2, RP33, U5-200KDApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-496