Learn More
Aviva Systems Biology Corp SNRP70 Antibody - N-terminal region (ARP40276_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40276_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SNRP70. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SNRP70. Alternate names: RPU1, Snp1, U1AP, U170K, U1RNP, RNPU1Z, SNRP70, U1-70KApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53185