Learn More
Aviva Systems Biology Corp SNRP70 Antibody - C-terminal region (ARP40439_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40439_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SNRP70. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SNRP70. Alternate names: RNPU1Z, RPU1, U170K, U1AP, U1RNP, Snp1, SNRP70, U1-70KApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53199