Learn More
Aviva Systems Biology Corp SNRPA Antibody - N-terminal region (ARP40530_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40530_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SNRPA. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SNRPA. Alternate names: Mud1, U1-A, U1AApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-284-879