Learn More
Aviva Systems Biology Corp SNRPB Antibody - N-terminal region (ARP40444_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40444_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SNRPB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SNRPB. Alternate names: COD, SNRPB1, SmB/SmB', snRNP-B, SmB/B', Sm-B/B'ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57457