Learn More
Aviva Systems Biology Corp SNRPD1 Antibody - N-terminal region (ARP40693_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40693_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SNRPD1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SNRPD1. Alternate names: SMD1, SNRPD, Sm-D1, HsT2456ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-53225