Learn More
ABclonal Technology SNRPD1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A144931000UL
Encompass_Preferred
This gene encodes a small nuclear ribonucleoprotein that belongs to the SNRNP core protein family. The protein may act as a charged protein scaffold to promote SNRNP assembly or strengthen SNRNP-SNRNP interactions through nonspecific electrostatic contacts with RNA. Two transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-06945