Learn More
Aviva Systems Biology Corp SNRPD2 Antibody - N-terminal region (ARP40532_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40532_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SNRPD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SNRPD2. Alternate names: SMD2, SNRPD1, Sm-D2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-57508