Learn More
Aviva Systems Biology Corp SNRPD2P2 ANTIBODY 50UG/50UL
SDP
Supplier:  Aviva Systems Biology Corp ARP57843_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC645339. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LOC645339. Alternate names: -ApplicationWB. Species reactivity: Human
Catalog No. 50-286-402