Learn More
Medchemexpress LLC HY-10008 10mg Medchemexpress, SNS-032 CAS:345627-80-7 Purity:>98%
SDP
Catalog No. 502028606
Encompass_Preferred
Medchemexpress, HY-10008 10mg SNS-032 CAS:345627-80-7 SNS-032 (BMS-387032) is a potent and selective inhibitor of CDK2, CDK7, and CDK9 with IC50s of 38 nM, 62 nM and 4 nM, respectively. SNS-032 has antitumor effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8606 Supplier Medchemexpress LLC Supplier No. HY1000810MG
Medchemexpress, HY-10008 10mg SNS-032 CAS:345627-80-7 SNS-032 (BMS-387032) is a potent and selective inhibitor of CDK2, CDK7, and CDK9 with IC50s of 38 nM, 62 nM and 4 nM, respectively. SNS-032 has antitumor effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.