Learn More
Selleck Chemical LLC SNS-032 (BMS-387032) 50mg 345627-80-7
SDP
Catalog No. 508749
Encompass_Preferred
SNS-032 (BMS-387032) has firstly been described as a selective inhibitor of CDK2 with IC50 of 48 nM in cell-free assays and is 10- and 20-fold selective over CDK1/CDK4. It is also found to be sensitive to CDK7/9 with IC50 of 62 nM/4 nM, with little effect on CDK6. SNS-032 (BMS-387032) induces apoptosis. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-874-9 Supplier Selleck Chemical LLC Supplier No. S114550MG
SNS-032 (BMS-387032) has firstly been described as a selective inhibitor of CDK2 with IC50 of 48 nM in cell-free assays and is 10- and 20-fold selective over CDK1/CDK4. It is also found to be sensitive to CDK7/9 with IC50 of 62 nM/4 nM, with little effect on CDK6. SNS-032 (BMS-387032) induces apoptosis. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.