Learn More
Medchemexpress LLC HY-12003 100mg , SNS-314 SNS-314 Mesylate CAS:1146618-41-8 Purity:98%
SDP
Catalog No. 501493401
Encompass_Preferred
Medchemexpress, HY-12003 100mg SNS-314 SNS-314 Mesylate CAS:1146618-41-8 Formula:C19H19ClN6O4S3 IC50: 9 nM (Aurora A), 31 nM (Aurora B), 6 nM (Aurora C) Purity:98% SNS-314 is a potent and selective aurora kinase inhibitor with IC 50 s of 9, 31, and 6 nM for aurora A, B and C, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3401 Supplier Medchemexpress LLC Supplier No. HY12003100MG
Medchemexpress, HY-12003 100mg SNS-314 SNS-314 Mesylate CAS:1146618-41-8 Formula:C19H19ClN6O4S3 IC50: 9 nM (Aurora A), 31 nM (Aurora B), 6 nM (Aurora C) Purity:98% SNS-314 is a potent and selective aurora kinase inhibitor with IC 50 s of 9, 31, and 6 nM for aurora A, B and C, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.