Learn More
Selleck Chemical LLC SNS-314 Mesylate
SDP
Catalog No. 5000001234
Encompass_Preferred
SNS-314 Mesylate is a potent and selective inhibitor of Aurora A Aurora B and Aurora C with IC50 of 9 nM 31 nM and 3 nM respectively and less potent to Trk A/B Flt4 Fms Axl c-Raf and DDR2
Catalog No. 50-000-01234 Supplier Selleck Chemical LLC Supplier No. S8699100MG
SNS-314 Mesylate is a potent and selective inhibitor of Aurora A Aurora B and Aurora C with IC50 of 9 nM 31 nM and 3 nM respectively and less potent to Trk A/B Flt4 Fms Axl c-Raf and DDR2