Learn More
Medchemexpress LLC Sns-314 mesylate | 1146618-41-8 | ≥98.0% | 25 MG
SDP
Catalog No. 5000234089
Encompass_Preferred
SNS-314 mesylate is the mesylate salt of SNS-314, a potent pan-Aurora kinase inhibitor active against Aurora A, B, and C used in preclinical research to investigate mitotic regulation and antineoplastic mechanisms.
  • Potent inhibition of Aurora A, B, and C (IC50 ≈ 9, 31, and 6 nM)
  • High purity: ≥98%
  • Powder form, soluble in DMSO for screening and formulation
  • Typical storage: -20 °C to preserve stability
  • Provided as mesylate salt; formula C18H15ClN6OS2 • CH3SO3H, formula weight 527.0
Catalog No. 50-002-34089 Supplier Medchemexpress LLC Supplier No. HY1200325MG
Add to Cart
Add to Cart
SNS-314 mesylate is the mesylate salt of SNS-314, a potent pan-Aurora kinase inhibitor active against Aurora A, B, and C used in preclinical research to investigate mitotic regulation and antineoplastic mechanisms.
  • Potent inhibition of Aurora A, B, and C (IC50 ≈ 9, 31, and 6 nM)
  • High purity: ≥98%
  • Powder form, soluble in DMSO for screening and formulation
  • Typical storage: -20 °C to preserve stability
  • Provided as mesylate salt; formula C18H15ClN6OS2 • CH3SO3H, formula weight 527.0