Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SNS-314 mesylate | 1146618-41-8 | >98.0% | 50 MG
SDP

Catalog No. 5000233315
Encompass_Preferred

SNS-314 mesylate is a potent pan-Aurora kinase inhibitor for preclinical research that inhibits Aurora A, B, and C at low nanomolar concentrations.

  • Potent inhibition of Aurora A, B, and C (IC50: Aurora A 9 nM; Aurora B 31 nM; Aurora C 6 nM).
  • Supplied as the mesylate salt for improved handling and stability.
  • Molecular formula C18H15ClN6OS2 · CH4O3S, molecular weight 527.04 g·mol-1.
  • High purity, typically >98.0% by HPLC.
  • Intended for in vitro research applications in cell cycle and oncology studies.

Catalog No. 50-002-33315 Supplier Medchemexpress LLC Supplier No. HY1200350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SNS-314 mesylate is a potent pan-Aurora kinase inhibitor for preclinical research that inhibits Aurora A, B, and C at low nanomolar concentrations.

  • Potent inhibition of Aurora A, B, and C (IC50: Aurora A 9 nM; Aurora B 31 nM; Aurora C 6 nM).
  • Supplied as the mesylate salt for improved handling and stability.
  • Molecular formula C18H15ClN6OS2 · CH4O3S, molecular weight 527.04 g·mol-1.
  • High purity, typically >98.0% by HPLC.
  • Intended for in vitro research applications in cell cycle and oncology studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.