Learn More
Medchemexpress LLC SNS-314 mesylate | 1146618-41-8 | 99.5% | 1 ML
SDP
Catalog No. 5000232445
Encompass_Preferred
SNS-314 mesylate is a potent, selective small-molecule inhibitor of Aurora kinases A, B, and C, with reported IC50 values of approximately 9 nM, 31 nM, and 6 nM, respectively. It is supplied for research use in solid form and as a ready-to-use 10 mM solution in DMSO.
  • Potent inhibition of Aurora A, B, and C (IC50: ~9, 31, and 6 nM)
  • High reported purity suitable for biochemical assays
  • Available as solid and as 10 mM solution in DMSO for convenience
  • Stable when stored sealed at recommended temperatures
  • White to light yellow solid, easy to handle
Catalog No. 50-002-32445 Supplier Medchemexpress LLC Supplier No. HY1200310MM 1ML
Add to Cart
Add to Cart
SNS-314 mesylate is a potent, selective small-molecule inhibitor of Aurora kinases A, B, and C, with reported IC50 values of approximately 9 nM, 31 nM, and 6 nM, respectively. It is supplied for research use in solid form and as a ready-to-use 10 mM solution in DMSO.
  • Potent inhibition of Aurora A, B, and C (IC50: ~9, 31, and 6 nM)
  • High reported purity suitable for biochemical assays
  • Available as solid and as 10 mM solution in DMSO for convenience
  • Stable when stored sealed at recommended temperatures
  • White to light yellow solid, easy to handle