Learn More
Selleck Chemical LLC SNS-314 5mg 1057249-41-8
SDP
Catalog No. 507173
Encompass_Preferred
SNS-314 is a potent and selective inhibitor of Aurora A, Aurora B and Aurora C with IC50 of 9 nM, 31 nM, and 3 nM, respectively. It is less potent to Trk A/B, Flt4, Fms, Axl, c-Raf and DDR2. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-717-3 Supplier Selleck Chemical LLC Supplier No. S11545MG
SNS-314 is a potent and selective inhibitor of Aurora A, Aurora B and Aurora C with IC50 of 9 nM, 31 nM, and 3 nM, respectively. It is less potent to Trk A/B, Flt4, Fms, Axl, c-Raf and DDR2. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.