Learn More
Cayman Chemical SNS-314 (mesylate), Purity: ≥98%, CAS: 1146618-41-8, 10 mg
SDP
Catalog No. 502971887
Encompass_Preferred
A pan-Aurora kinase inhibitor (IC50s = 9, 31, and 3.4 nM for Aurora A, B, and C, respectively); inhibits TRKB, TRKA, FLT4, FMS, DDR2, AXL, and C-RAF (IC50s = 5-100 nM); inhibits Aurora B phosphorylation of histone H3 in HCT116 human colorectal carcinoma cells in vitro (EC50 = <16 nM) and in a mouse xenograft model at a dose of 50 mg/kg; inhibits tumor growth in A2780 ovarian, A375 melanoma, H1299 lung, MDA-MB-231 breast, and PC3 prostate cancer mouse xenograft models
Catalog No. 50-297-1887 Supplier Cayman Chemical Supplier No. 1497410 MG
A pan-Aurora kinase inhibitor (IC50s = 9, 31, and 3.4 nM for Aurora A, B, and C, respectively); inhibits TRKB, TRKA, FLT4, FMS, DDR2, AXL, and C-RAF (IC50s = 5-100 nM); inhibits Aurora B phosphorylation of histone H3 in HCT116 human colorectal carcinoma cells in vitro (EC50 = <16 nM) and in a mouse xenograft model at a dose of 50 mg/kg; inhibits tumor growth in A2780 ovarian, A375 melanoma, H1299 lung, MDA-MB-231 breast, and PC3 prostate cancer mouse xenograft models