Learn More
Medchemexpress LLC HY-11029 10mg Medchemexpress, SNT-207707 CAS:1064662-40-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1102910MG
Encompass_Preferred
Medchemexpress, HY-11029 10mg SNT-207707 CAS:1064662-40-3 SNT-207707 is a selective, potent and orally active melanocortin MC-4 receptor antagonist with an IC50 of 8 nM (binding) and 5 nM (function) on the MC-4 receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8788