Learn More
Medchemexpress LLC Snt-207858 (free base) | 1104662-66-9 | 99.7% | 616.62 | 1 ML
SDP
Catalog No. 5000343936
Encompass_Preferred
SNT-207858 free base is a selective, blood-brain barrier penetrating, potent, and orally active melanocortin-4 (MC-4) receptor antagonist. It demonstrates high affinity and selectivity for the MC-4 receptor, with an IC50 of 22 nM (binding) and 11 nM (function), showing 170-fold selectivity over MC-3 and 40-fold over MC-5. In preclinical studies, SNT-207858 significantly reduced tumor-induced weight loss in mice.
  • Selective melanocortin-4 (MC-4) receptor antagonist
  • Blood-brain barrier penetrating
  • Orally active
  • High potency with IC50 of 22 nM (binding) and 11 nM (function) on MC-4 receptor
  • Shows 170-fold selectivity versus MC-3 and 40-fold selectivity versus MC-5
  • Significantly reduces tumor-induced weight loss in mice with C26 adenocarcinoma-induced cachexia
Catalog No. 50-003-43936 Supplier Medchemexpress LLC Supplier No. HY11030A100MG
SNT-207858 free base is a selective, blood-brain barrier penetrating, potent, and orally active melanocortin-4 (MC-4) receptor antagonist. It demonstrates high affinity and selectivity for the MC-4 receptor, with an IC50 of 22 nM (binding) and 11 nM (function), showing 170-fold selectivity over MC-3 and 40-fold over MC-5. In preclinical studies, SNT-207858 significantly reduced tumor-induced weight loss in mice.
  • Selective melanocortin-4 (MC-4) receptor antagonist
  • Blood-brain barrier penetrating
  • Orally active
  • High potency with IC50 of 22 nM (binding) and 11 nM (function) on MC-4 receptor
  • Shows 170-fold selectivity versus MC-3 and 40-fold selectivity versus MC-5
  • Significantly reduces tumor-induced weight loss in mice with C26 adenocarcinoma-induced cachexia