Learn More
GLATFELTER SONTARA OLD HICKORY INC SNTARAEC WH FLT 12X12 WPE BULK
Catalog No. NC9009065
Encompass_Preferred
Durable Creped Finish Binder Free Excellent Tear Strength Solvent Resistant Poly Pack Wipes : 100 Per Pack Quantity : 10 Packs Per Case Type : Flat Polybagged Color: White
This item is not returnable. View return policy
Catalog No. NC9009065 Supplier GLATFELTER SONTARA OLD HICKORY INC Supplier No. PR911
Durable Creped Finish Binder Free Excellent Tear Strength Solvent Resistant Poly Pack Wipes : 100 Per Pack Quantity : 10 Packs Per Case Type : Flat Polybagged Color: White