Learn More
Aviva Systems Biology Corp SNTB2 Antibody - middle region (ARP62913_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62913_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SNTB2. It was validated on Western Blot. . Alternate names: D16S2531E, EST25263, SNT2B2, SNT3, SNTLApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-804