Learn More
Aviva Systems Biology Corp Sntg1 Antibody - N-terminal region (ARP57299_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57299_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Sntg1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 4933426D16Rik, G1SYN, SYN4ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-295