Learn More
Med Vet International Sentinel Spectrum Blue for Dogs, 50.1-100 lbs., 6/card, 5 cards/box chews
Catalog No. 503045452
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Sentinel Spectrum beef and bacon flavored soft chewables provide the broadest spectrum of protection against heartworm available on the market today. Indicated to provide treatment and control of adult roundworms, hookworms, whipworms and adult tapeworms as well as control of fleas. Contains Milbemycin Oxime, Praziquantel (offering treatment and control of tapeworms), and Lufenuron (stopping fleas before they become adults, thereby breaking the flea lifecycle). Pillow-shaped chewables facilitate dosing. Each chewable is cured, cut and packaged in a protective blister pack to preserve freshness. For dogs and puppies six weeks of age and older.
Catalog No. 50-304-5452 Supplier Med Vet International Supplier No. RXSENSPECBL6
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Sentinel Spectrum beef and bacon flavored soft chewables provide the broadest spectrum of protection against heartworm available on the market today. Indicated to provide treatment and control of adult roundworms, hookworms, whipworms and adult tapeworms as well as control of fleas. Contains Milbemycin Oxime, Praziquantel (offering treatment and control of tapeworms), and Lufenuron (stopping fleas before they become adults, thereby breaking the flea lifecycle). Pillow-shaped chewables facilitate dosing. Each chewable is cured, cut and packaged in a protective blister pack to preserve freshness. For dogs and puppies six weeks of age and older.