Learn More
ATCC SNU-16; Gastric Carcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL5974
Encompass
SNU-16 is a cell line exhibiting epithelial morphology that was isolated in 1987 from ascites derived from a 33-year-old, female, Asian, stomach cancer patient prior to chemotherapy. Use these cells in cancer research.
Catalog No. 50-238-5039