Learn More
Grainger Starrett Snug Assembly, 5/16 in. Post Hole, 3/32 to 1/4 in. Capacity, 1 ea.
Catalog No. 502787657
Encompass_Preferred
This snug assembly is a precision machining accessory designed for versatile use with height transfer gauges, surface gauges, and magnetic bases. It features a robust design to securely hold components, facilitating accurate measurements and reliable tool positioning.
  • Fits a 5/16 in. post hole
  • Provides a holding capacity of 3/32 in. to 1/4 in.
  • Compatible with swivel post assemblies
  • Manufactured in the USA
Catalog No. 50-278-7657 Supplier Grainger Supplier No. 197R42
This snug assembly is a precision machining accessory designed for versatile use with height transfer gauges, surface gauges, and magnetic bases. It features a robust design to securely hold components, facilitating accurate measurements and reliable tool positioning.
  • Fits a 5/16 in. post hole
  • Provides a holding capacity of 3/32 in. to 1/4 in.
  • Compatible with swivel post assemblies
  • Manufactured in the USA