Learn More
Aviva Systems Biology Corp SNUPN Antibody - middle region (ARP40598_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40598_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SNUPN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SNUPN. Alternate names: KPNBL, RNUT1, Snurportin1ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-57540