Learn More
Medchemexpress LLC SNX-0723 | 1073969-18-2 | 99.1% | 399.46 g·mol^-1 | C22H26FN3O3 | 50 MG
SDP
Catalog No. 5000227344
Encompass_Preferred
SNX-0723 is a brain-permeable small-molecule Hsp90 inhibitor reported to have anti-Plasmodium and antitumor activity. It is supplied for research use in high-purity solid and solution formats for in vitro and in vivo studies.
  • Potent Hsp90 inhibitor with reported biological activity.
  • Reported purity ~99.1% suitable for research applications.
  • Molecular weight 399.46 g·mol⁻¹ and formula C22H26FN3O3.
  • Available as solid powder and as DMSO solution for flexibility.
  • Stored under recommended low-temperature conditions for stability.
Catalog No. 50-002-27344 Supplier Medchemexpress LLC Supplier No. HY11904650MG
SNX-0723 is a brain-permeable small-molecule Hsp90 inhibitor reported to have anti-Plasmodium and antitumor activity. It is supplied for research use in high-purity solid and solution formats for in vitro and in vivo studies.
  • Potent Hsp90 inhibitor with reported biological activity.
  • Reported purity ~99.1% suitable for research applications.
  • Molecular weight 399.46 g·mol⁻¹ and formula C22H26FN3O3.
  • Available as solid powder and as DMSO solution for flexibility.
  • Stored under recommended low-temperature conditions for stability.