Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SNX-0723 | 1073969-18-2 | 99.1% | 399.46 g/mol | C22H26FN3O3 | 10 MG
SDP

Catalog No. 5000228943
Encompass_Preferred

SNX-0723 is a small-molecule Hsp90 inhibitor used in research to investigate Hsp90 biology and anti-plasmodial activity. It is supplied as a solid for in vitro and in vivo applications.

  • Potent Hsp90 inhibitor active against human and Plasmodium Hsp90.
  • High purity (≈99.1%).
  • Molecular weight 399.46 g/mol; formula C22H26FN3O3.
  • Off-white to light yellow solid; supplied in 10 mg amount.
  • Soluble in DMSO (~20.83 mg/mL); in vivo formulation protocols available.
  • Powder storage: -20°C for long-term stability; solution storage at -80°C recommended.

Catalog No. 50-002-28943 Supplier Medchemexpress LLC Supplier No. HY11904610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SNX-0723 is a small-molecule Hsp90 inhibitor used in research to investigate Hsp90 biology and anti-plasmodial activity. It is supplied as a solid for in vitro and in vivo applications.

  • Potent Hsp90 inhibitor active against human and Plasmodium Hsp90.
  • High purity (≈99.1%).
  • Molecular weight 399.46 g/mol; formula C22H26FN3O3.
  • Off-white to light yellow solid; supplied in 10 mg amount.
  • Soluble in DMSO (~20.83 mg/mL); in vivo formulation protocols available.
  • Powder storage: -20°C for long-term stability; solution storage at -80°C recommended.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.