Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SNX-0723 | 1073969-18-2 | 99.1% | 399.46 g/mol | C22H26FN3O3 | 25 MG
SDP

Catalog No. 5000228347
Encompass_Preferred

This small-molecule Hsp90 inhibitor is supplied for biochemical and cellular research applications. It exhibits potent binding to human and Plasmodium Hsp90 and is provided as a high-purity reagent for in vitro studies.

  • High purity of 99.1% supports reproducible results.
  • Soluble in DMSO for straightforward preparation of stock solutions.
  • Molecular formula C22H26FN3O3 and molecular weight 399.46 g/mol aid in calculations and dosing.
  • Available in small research-friendly quantities for screening and assay development.

Catalog No. 50-002-28347 Supplier Medchemexpress LLC Supplier No. HY11904625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This small-molecule Hsp90 inhibitor is supplied for biochemical and cellular research applications. It exhibits potent binding to human and Plasmodium Hsp90 and is provided as a high-purity reagent for in vitro studies.

  • High purity of 99.1% supports reproducible results.
  • Soluble in DMSO for straightforward preparation of stock solutions.
  • Molecular formula C22H26FN3O3 and molecular weight 399.46 g/mol aid in calculations and dosing.
  • Available in small research-friendly quantities for screening and assay development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.