Learn More
Medchemexpress LLC Snx-2112 | 908112-43-6 | 99.5% | 464.48 g/mol | C23H27F3N4O3 | 200 MG
SDP
Catalog No. 5000246941
Encompass_Preferred
SNX-2112 is an Hsp90 inhibitor research compound used to study Hsp90-mediated signaling and cancer cell biology. It binds Hsp90 isoforms with low-nanomolar affinity and induces degradation of client proteins, making it suitable for biochemical and cellular assays.
  • High purity (99.5%) appropriate for sensitive assays.
  • Molecular weight 464.48 g/mol.
  • White to off-white solid, easy to weigh and dissolve.
  • Stable when stored as a powder at -20°C for long-term storage.
  • Supplied in a 200 MG quantity for laboratory use.
Catalog No. 50-002-46941 Supplier Medchemexpress LLC Supplier No. HY10214200MG
SNX-2112 is an Hsp90 inhibitor research compound used to study Hsp90-mediated signaling and cancer cell biology. It binds Hsp90 isoforms with low-nanomolar affinity and induces degradation of client proteins, making it suitable for biochemical and cellular assays.
  • High purity (99.5%) appropriate for sensitive assays.
  • Molecular weight 464.48 g/mol.
  • White to off-white solid, easy to weigh and dissolve.
  • Stable when stored as a powder at -20°C for long-term storage.
  • Supplied in a 200 MG quantity for laboratory use.