Learn More
Selleck Chemical LLC SNX-2112 (PF-04928473) 5mg 908112-43-6
SDP
Catalog No. 501362689
Encompass_Preferred
SNX-2112 (PF-04928473) selectively binds to the ATP pocket of HSP90? and HSP90? with Ka of 30 nM and 30 nM, uniformly more potent than 17-AAG. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2689 Supplier Selleck Chemical LLC Supplier No. S26395MG
SNX-2112 (PF-04928473) selectively binds to the ATP pocket of HSP90? and HSP90? with Ka of 30 nM and 30 nM, uniformly more potent than 17-AAG. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.