Learn More
Aviva Systems Biology Corp SNX1 Antibody - N-terminal region (ARP64037_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64037_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SNX1. It was validated on Western Blot. . Alternate names: HsT17379, MGC8664, SNX1A, Vps5, VPS5ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-151