Learn More
Aviva Systems Biology Corp Snx10 Antibody - middle region (ARP54936_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54936_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Snx10. It was validated on Western Blot. . Alternate names: 2410004M09RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-980