Learn More
Aviva Systems Biology Corp SNX11 Antibody - middle region (ARP55482_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55482_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SNX11. It was validated on Western Blot. . Alternate names: MGC111019ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-286-043