Learn More
Aviva Systems Biology Corp SNX12 Antibody - middle region (ARP62915_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62915_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SNX12. It was validated on Western Blot. . Alternate names: MGC118982, MGC118983ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-805