Learn More
Aviva Systems Biology Corp SNX21 Antibody - N-terminal region (ARP62918_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62918_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SNX21. It was validated on Western Blot. . Alternate names: C20orf161, MGC29895, PP3993, SNX-L, dJ337O18.4ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit
Catalog No. 50-287-806