Learn More
TARGETMOL CHEMICALS INC SNX2112 5MG
SDP
Catalog No. 502579445
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SNX2112 (PF 04928473) is an orally active Hsp90 inhibitor with a Kd of 16 nM. purity: 99%
Catalog No. 50-257-9445 Supplier TARGETMOL CHEMICALS INC Supplier No. T63055MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SNX2112 (PF 04928473) is an orally active Hsp90 inhibitor with a Kd of 16 nM. purity: 99%