Learn More
Aviva Systems Biology Corp SNX5 ANTIBODY 50UG/50UL
SDP
Supplier:  Aviva Systems Biology Corp ARP55049_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Snx5. It was validated on Western Blot. . Alternate names: RP23-35E16.2, 0910001N05Rik, 1810032P22Rik, AU019504, D2Ertd52eApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-994