Learn More
Aviva Systems Biology Corp SNX7 Antibody - middle region (ARP56795_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56795_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SNX7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SNX7. Alternate names: DKFZP564F052, MGC8717ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61992