Learn More
Aviva Systems Biology Corp SNX8 Antibody - C-terminal region (ARP62716_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62716_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SNX8. It was validated on Western Blot. . Alternate names: Mvp1ApplicationWB. Species reactivity: Guinea Pig, Human, Mouse, Rat
Catalog No. 50-287-752