Learn More
MilliporeSigma™ Supelco™ Snyder Columns
Catalog No. 111004538 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Description:
2-Ball Design
3-Ball Design
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
11-100-4538 3-Ball Design
11-100-4533 2-Ball Design
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-4538 Supplier MilliporeSigma™ Supelco™ Supplier No. 64705U
Add to Cart
Add to Cart
Utilize separate chambers to promote multiple condensations and vaporizations during the distillation process, leading to a more efficient separation
TRUSTED_SUSTAINABILITY
Specifications
Connector Types Threaded Joint
Description 3-Ball Design
Quantity 1 Ea.
For Use With (Application) Condensations and Vaporizations
Type Snyder Column