Learn More
Sklar Instruments Instrument Soaking Tray, Sklar, Medium. Three piece set includes tray, strainer and lid. Clear Plastic. For Storage/Transportation Only. 7 in. x 13 in. x 4 in. Deep. PK/3.
SDP
Catalog No. 501437180 Shop All Sklar Instruments Products
Encompass_Preferred
Instrument Soaking Tray, Sklar, Medium. Three piece set includes tray, strainer and lid. Clear Plastic. For Storage/Transportation Only. 7 in. x 13 in. x 4 in. Deep. Sklar's Instrument Soaking Trays are containers used for sterilizing instruments by soaking in cleaning solution. They are three piece sets that includes a bottom, perforated tray, and lid. They are available in various sizes and styles.
This item is not returnable. View return policy
Catalog No. 50-143-7180 Supplier Sklar Instruments Supplier No. 103054
Instrument Soaking Tray, Sklar, Medium. Three piece set includes tray, strainer and lid. Clear Plastic. For Storage/Transportation Only. 7 in. x 13 in. x 4 in. Deep. Sklar's Instrument Soaking Trays are containers used for sterilizing instruments by soaking in cleaning solution. They are three piece sets that includes a bottom, perforated tray, and lid. They are available in various sizes and styles.